Learn More
Grainger Wire Rope Locking Stud RH Thread 5/16 in
Catalog No. 502765661
Encompass_Preferred
The Grainger 16X735 is a 16-inch diameter, 735 CFM, direct drive, aluminum exhaust fan. It features a 1/3 HP, 115V motor with thermal overload protection and sealed ball bearings. The fan has a maximum speed of 1525 RPM. It is designed for exhaust applications and is UL/cUL listed. The housing is constructed of aluminum with a gray powder coat finish.
Catalog No. 50-276-5661 Supplier Grainger Supplier No. 16X735
The Grainger 16X735 is a 16-inch diameter, 735 CFM, direct drive, aluminum exhaust fan. It features a 1/3 HP, 115V motor with thermal overload protection and sealed ball bearings. The fan has a maximum speed of 1525 RPM. It is designed for exhaust applications and is UL/cUL listed. The housing is constructed of aluminum with a gray powder coat finish.