Learn More
Grainger Wire Rope Locking Stud RH Thread 1/8 in
Catalog No. 502765663
Encompass_Preferred
The Loos Wire Rope Locking Stud 1/8-16 is a wire rope stud designed for use with 1/8" wire rope. It is made from carbon steel and zinc plated for corrosion resistance. This stud has a working load limit of 350 lbs and ultimate strength of 700 lbs. It can be used for a variety of wire rope termination applications. The stud measures 0.31" tall with a thread diameter of 0.25". It comes in a package of 100 pieces. This is an economical wire rope stud for light duty wire rope applications.
Catalog No. 50-276-5663 Supplier Grainger Supplier No. 16X719
The Loos Wire Rope Locking Stud 1/8-16 is a wire rope stud designed for use with 1/8" wire rope. It is made from carbon steel and zinc plated for corrosion resistance. This stud has a working load limit of 350 lbs and ultimate strength of 700 lbs. It can be used for a variety of wire rope termination applications. The stud measures 0.31" tall with a thread diameter of 0.25". It comes in a package of 100 pieces. This is an economical wire rope stud for light duty wire rope applications.