Learn More
Grainger Wire Rope Locking Stud RH Thread 5/32 in
Catalog No. 502765685
Encompass_Preferred
The Grainger Approved Wire Rope Locking Stud is a 1/2" diameter wire rope stud with a working load limit of 1,250 lbs. It is made from drop forged carbon steel and zinc plated for corrosion resistance. The stud has a threaded stud on one end to accept a nut and a socket on the other end to accept a swaged wire rope fitting. It can be used for making wire rope connections in rigging applications. The stud meets ASME B30.26 specifications.
Catalog No. 50-276-5685 Supplier Grainger Supplier No. 16X723
The Grainger Approved Wire Rope Locking Stud is a 1/2" diameter wire rope stud with a working load limit of 1,250 lbs. It is made from drop forged carbon steel and zinc plated for corrosion resistance. The stud has a threaded stud on one end to accept a nut and a socket on the other end to accept a swaged wire rope fitting. It can be used for making wire rope connections in rigging applications. The stud meets ASME B30.26 specifications.