Learn More
Grainger Wire Rope Locking Stud RH Thread 5/32 in
Catalog No. 502768195
Encompass_Preferred
The Grainger 16X721 Wire Rope Locking Stud has a right hand thread, is made of alloy steel, and has a working load limit of 1,250 lbs. It is designed for terminating wire rope by inserting it through the eye and swaging. The stud's dimensions are 0.5" diameter x 1.75" length. It can be used in rigging applications. The stud should be installed using the proper swaging tools for secure wire rope termination. Proper installation helps prevent wire rope slippage and failure.
Catalog No. 50-276-8195 Supplier Grainger Supplier No. 16X721
The Grainger 16X721 Wire Rope Locking Stud has a right hand thread, is made of alloy steel, and has a working load limit of 1,250 lbs. It is designed for terminating wire rope by inserting it through the eye and swaging. The stud's dimensions are 0.5" diameter x 1.75" length. It can be used in rigging applications. The stud should be installed using the proper swaging tools for secure wire rope termination. Proper installation helps prevent wire rope slippage and failure.