Learn More
Grainger Wire Rope Locking Stud RH Thread 1/16 in
Catalog No. 502768236
Encompass_Preferred
The Loos Wire Rope Locking Stud is a 1/16" diameter stud designed for wire rope applications. It is made from grade 5 titanium with a tensile strength of 120,000 psi. The stud has a head diameter of 3/16" and an installed height of 1/4". It can be used with 1x7 and 7x7 wire rope constructions. The stud is installed using a standard stud gun and features a locking collar to prevent loosening under vibration. It provides a secure termination point for wire rope in a variety of industrial lifting, rigging and cable railing applications.
Catalog No. 50-276-8236 Supplier Grainger Supplier No. 16X711
The Loos Wire Rope Locking Stud is a 1/16" diameter stud designed for wire rope applications. It is made from grade 5 titanium with a tensile strength of 120,000 psi. The stud has a head diameter of 3/16" and an installed height of 1/4". It can be used with 1x7 and 7x7 wire rope constructions. The stud is installed using a standard stud gun and features a locking collar to prevent loosening under vibration. It provides a secure termination point for wire rope in a variety of industrial lifting, rigging and cable railing applications.