Learn More
Grainger Wire Rope Locking Stud RH Thread 3/8 in
Catalog No. 502765115
Encompass_Preferred
The Grainger Approved Wire Rope Locking Stud is a 16mm stud designed for wire rope applications. It is made from grade 8 zinc plated steel and features a head design that prevents spinning. The stud has a minimum ultimate strength of 150,000 psi. It can be used with 1/4 to 3/8 inch wire rope. The stud's dimensions are 0.63 x 0.63 x 1.5 inches. It weighs 0.05 pounds. This wire rope stud provides a secure termination point and its anti-spin head allows for easy installation. The high strength steel construction makes it suitable for demanding wire rope applications.
Catalog No. 50-276-5115 Supplier Grainger Supplier No. 16X737
The Grainger Approved Wire Rope Locking Stud is a 16mm stud designed for wire rope applications. It is made from grade 8 zinc plated steel and features a head design that prevents spinning. The stud has a minimum ultimate strength of 150,000 psi. It can be used with 1/4 to 3/8 inch wire rope. The stud's dimensions are 0.63 x 0.63 x 1.5 inches. It weighs 0.05 pounds. This wire rope stud provides a secure termination point and its anti-spin head allows for easy installation. The high strength steel construction makes it suitable for demanding wire rope applications.