Learn More
Grainger Wire Rope Oval Sleeve 1/16 in SS
Catalog No. 502766993
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of galvanized steel. It has an inside diameter of 16mm and is designed to fit 6x19 wire rope. The sleeve is 26mm long and features a closed seam. It can be used to help protect wire rope from wear and tear. The sleeve is sold individually.
Catalog No. 50-276-6993 Supplier Grainger Supplier No. 16X826
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of galvanized steel. It has an inside diameter of 16mm and is designed to fit 6x19 wire rope. The sleeve is 26mm long and features a closed seam. It can be used to help protect wire rope from wear and tear. The sleeve is sold individually.