Learn More
Grainger Wire Rope Oval Sleeve 1/16 in SS
Catalog No. 502765516
Encompass_Preferred
The Grainger 16X827 is a 16-inch diameter, 827 CFM, 3-speed, direct drive exhaust fan. It features a thermally protected motor and is constructed of galvanized steel with a gray powder coat finish. This exhaust fan is designed for wall mounting and has a sound rating of 7.4 sones. It operates on 120V power and weighs approximately 35 lbs. The 16X827 exhaust fan from Grainger provides high airflow for industrial and commercial ventilation needs.
Catalog No. 50-276-5516 Supplier Grainger Supplier No. 16X827
The Grainger 16X827 is a 16-inch diameter, 827 CFM, 3-speed, direct drive exhaust fan. It features a thermally protected motor and is constructed of galvanized steel with a gray powder coat finish. This exhaust fan is designed for wall mounting and has a sound rating of 7.4 sones. It operates on 120V power and weighs approximately 35 lbs. The 16X827 exhaust fan from Grainger provides high airflow for industrial and commercial ventilation needs.