Learn More
Grainger Wire Rope Oval Sleeve 1/8 in SS
Catalog No. 502768229
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of drop forged steel. It has a working load limit of 3 tons and is designed to fit 1/4" wire rope. The sleeve measures 1-1/4" long x 15/16" wide x 13/16" high. It can be used for various rigging and hoisting applications to protect and reinforce the ends of wire rope. The drop forged construction ensures durability and strength. Overall, this is a heavy-duty wire rope end fitting for securing and protecting 1/4" wire rope in rigging systems.
Catalog No. 50-276-8229 Supplier Grainger Supplier No. 16X828
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of drop forged steel. It has a working load limit of 3 tons and is designed to fit 1/4" wire rope. The sleeve measures 1-1/4" long x 15/16" wide x 13/16" high. It can be used for various rigging and hoisting applications to protect and reinforce the ends of wire rope. The drop forged construction ensures durability and strength. Overall, this is a heavy-duty wire rope end fitting for securing and protecting 1/4" wire rope in rigging systems.