Learn More
Grainger Wire Rope Oval Sleeve 1/8 in SS
Catalog No. 502767922
Encompass_Preferred
The Grainger 1/8" Stainless Steel Wire Rope Oval Sleeve is made from 316 stainless steel for corrosion resistance. It has a working load limit of 110 lbs and is designed for 1/8" 7x7 or 7x19 wire rope. The sleeve measures 1-1/4" long x 3/4" wide x 1/4" thick. This wire rope sleeve can be used for various rigging applications to help protect the rope from wear and tear. The oval shape distributes load evenly. Overall this is a high quality stainless steel wire rope sleeve for 1/8" diameter wire rope.
Catalog No. 50-276-7922 Supplier Grainger Supplier No. 16X829
The Grainger 1/8" Stainless Steel Wire Rope Oval Sleeve is made from 316 stainless steel for corrosion resistance. It has a working load limit of 110 lbs and is designed for 1/8" 7x7 or 7x19 wire rope. The sleeve measures 1-1/4" long x 3/4" wide x 1/4" thick. This wire rope sleeve can be used for various rigging applications to help protect the rope from wear and tear. The oval shape distributes load evenly. Overall this is a high quality stainless steel wire rope sleeve for 1/8" diameter wire rope.