Learn More
Grainger Wire Rope Oval Sleeve 3/16 in SS
Catalog No. 502766989
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of galvanized steel. It is designed to fit 1/4" wire rope and has an inside width of 0.27", an inside depth of 0.39", and a length of 1.5". This wire rope sleeve can be used to help secure the ends of wire rope to make a loop or connect two sections of wire rope together. The galvanized steel construction helps prevent corrosion. Overall, this is a basic wire rope sleeve for use with 1/4" galvanized steel wire rope in various applications.
Catalog No. 50-276-6989 Supplier Grainger Supplier No. 16X831
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of galvanized steel. It is designed to fit 1/4" wire rope and has an inside width of 0.27", an inside depth of 0.39", and a length of 1.5". This wire rope sleeve can be used to help secure the ends of wire rope to make a loop or connect two sections of wire rope together. The galvanized steel construction helps prevent corrosion. Overall, this is a basic wire rope sleeve for use with 1/4" galvanized steel wire rope in various applications.