Learn More
Grainger Wire Rope Oval Sleeve 3/16 in SS
Catalog No. 502766799
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of drop forged steel. It has a working load limit of 3 tons and is designed for 1/4" wire rope. The sleeve measures 1-1/4" long x 15/16" wide x 13/16" high. It can be used for wire rope repair and maintenance in industrial settings. The sleeve is zinc plated for corrosion resistance. This wire rope sleeve provides a secure connection when used properly with the appropriate size wire rope.
Catalog No. 50-276-6799 Supplier Grainger Supplier No. 16X830
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of drop forged steel. It has a working load limit of 3 tons and is designed for 1/4" wire rope. The sleeve measures 1-1/4" long x 15/16" wide x 13/16" high. It can be used for wire rope repair and maintenance in industrial settings. The sleeve is zinc plated for corrosion resistance. This wire rope sleeve provides a secure connection when used properly with the appropriate size wire rope.