Learn More
Grainger Lift-All Wire Rope Sling, Steel, 6 x 19, 1/2 in., 6 ft., 1 ea
Catalog No. 502794280
Encompass_Preferred
This eye-and-eye wire rope sling is designed for secure rigging and lifting applications. Constructed from durable steel with an Independent Wire Rope Core (IWRC), this 1/2 in. diameter sling features 6 x 19 construction for a balance of flexibility and abrasion resistance. Each sling utilizes the Flemish Eye Splice technique, which provides reserve strength beyond that of the swaged sleeve alone. Suitable for vertical, choker, and basket hitch configurations, this product meets OSHA 1910.184 and ASME B30.9 standards.
  • 6 x 19 classification with EIPS grade steel
  • Independent wire rope core for enhanced durability
  • Abrasion-resistant design for demanding environments
  • Operating temperature range from -60 °F to +400 °F
  • Rated vertical lifting capacity of 5,000 lb.
  • Uncoated finish
Catalog No. 50-279-4280 Supplier Grainger Supplier No. 1A593
This eye-and-eye wire rope sling is designed for secure rigging and lifting applications. Constructed from durable steel with an Independent Wire Rope Core (IWRC), this 1/2 in. diameter sling features 6 x 19 construction for a balance of flexibility and abrasion resistance. Each sling utilizes the Flemish Eye Splice technique, which provides reserve strength beyond that of the swaged sleeve alone. Suitable for vertical, choker, and basket hitch configurations, this product meets OSHA 1910.184 and ASME B30.9 standards.
  • 6 x 19 classification with EIPS grade steel
  • Independent wire rope core for enhanced durability
  • Abrasion-resistant design for demanding environments
  • Operating temperature range from -60 °F to +400 °F
  • Rated vertical lifting capacity of 5,000 lb.
  • Uncoated finish