Learn More
Grainger Wire Rope Stop Sleeve 1/16 in SS
Catalog No. 502765676
Encompass_Preferred
The Loos Wire Rope Stop Sleeve Stop is a wire rope end fitting designed to prevent fraying and unraveling of wire rope ends. It is made from drop forged steel and has a U-bolt design that allows for easy installation. This wire rope stop can be used on 6x7 and 6x19 class wire rope and comes in sizes ranging from 1/4" to 1" diameter. It meets Fed Spec RR-C-271 Type I Class 1 and is RoHS compliant. The all steel construction provides durability while the U-bolt allows the stop to be securely tightened onto the wire rope end. Overall, this is a heavy duty wire rope stop that prevents fraying and unraveling to protect wire rope ends.
Catalog No. 50-276-5676 Supplier Grainger Supplier No. 16X634
The Loos Wire Rope Stop Sleeve Stop is a wire rope end fitting designed to prevent fraying and unraveling of wire rope ends. It is made from drop forged steel and has a U-bolt design that allows for easy installation. This wire rope stop can be used on 6x7 and 6x19 class wire rope and comes in sizes ranging from 1/4" to 1" diameter. It meets Fed Spec RR-C-271 Type I Class 1 and is RoHS compliant. The all steel construction provides durability while the U-bolt allows the stop to be securely tightened onto the wire rope end. Overall, this is a heavy duty wire rope stop that prevents fraying and unraveling to protect wire rope ends.