Learn More
Grainger Wire Rope Stop Sleeve 1/8 in SS
Catalog No. 502767976
Encompass_Preferred
The Loos Wire Rope Stop Sleeve Stop is a wire rope termination device designed to prevent fraying of wire rope ends. It is made from drop-forged steel and has a working load limit of 9,000 lbs. The sleeve accommodates 1/4"-5/8" diameter 7x7 and 7x19 wire rope. It is 3-1/8" long x 1-3/4" wide and weighs 0.60 lbs. The Loos sleeve stop meets military specification MIL-S-21433. It provides a finished look to wire rope ends in industrial lifting applications.
Catalog No. 50-276-7976 Supplier Grainger Supplier No. 16X635
The Loos Wire Rope Stop Sleeve Stop is a wire rope termination device designed to prevent fraying of wire rope ends. It is made from drop-forged steel and has a working load limit of 9,000 lbs. The sleeve accommodates 1/4"-5/8" diameter 7x7 and 7x19 wire rope. It is 3-1/8" long x 1-3/4" wide and weighs 0.60 lbs. The Loos sleeve stop meets military specification MIL-S-21433. It provides a finished look to wire rope ends in industrial lifting applications.