Learn More
Grainger Wire Rope Stop Sleeve 1/8 in SS
Catalog No. 502765677
Encompass_Preferred
The Loos Wire Rope Stop Sleeve Stop is a wire rope fitting designed to prevent fraying of wire rope ends. It is made from drop-forged steel and has a U-bolt design with serrated edges to grip the wire rope securely. This wire rope stop can be installed by sliding it over the end of the wire rope and tightening the U-bolt using a wrench. It is available in multiple sizes to accommodate wire rope diameters from 1/4" to 1-1/4". The working load limit ranges from 1,260 lbs to 23,760 lbs depending on the wire rope diameter. This wire rope end stop provides a finished look and helps protect against accidents caused by frayed wire rope ends.
Catalog No. 50-276-5677 Supplier Grainger Supplier No. 16X636
The Loos Wire Rope Stop Sleeve Stop is a wire rope fitting designed to prevent fraying of wire rope ends. It is made from drop-forged steel and has a U-bolt design with serrated edges to grip the wire rope securely. This wire rope stop can be installed by sliding it over the end of the wire rope and tightening the U-bolt using a wrench. It is available in multiple sizes to accommodate wire rope diameters from 1/4" to 1-1/4". The working load limit ranges from 1,260 lbs to 23,760 lbs depending on the wire rope diameter. This wire rope end stop provides a finished look and helps protect against accidents caused by frayed wire rope ends.