Learn More
Grainger Wire Rope Stop Sleeve 3/16 in SS
Catalog No. 502768141
Encompass_Preferred
The Loos Wire Rope Stop Sleeve Stop is a wire rope termination fitting designed to prevent fraying and unraveling of wire rope ends. It is made from drop forged steel and has a U-bolt design with nuts to clamp it onto the wire rope end. This stop sleeve is available in multiple sizes to accommodate wire rope diameters from 1/4" to 1" and safe working loads from 1,260 lbs to 15,000 lbs. The U-bolt opening sizes range from 0.31" to 1.06" to fit the different wire rope diameters. This is a heavy duty stop sleeve made in the USA that provides a secure end termination for wire rope.
Catalog No. 50-276-8141 Supplier Grainger Supplier No. 16X637
The Loos Wire Rope Stop Sleeve Stop is a wire rope termination fitting designed to prevent fraying and unraveling of wire rope ends. It is made from drop forged steel and has a U-bolt design with nuts to clamp it onto the wire rope end. This stop sleeve is available in multiple sizes to accommodate wire rope diameters from 1/4" to 1" and safe working loads from 1,260 lbs to 15,000 lbs. The U-bolt opening sizes range from 0.31" to 1.06" to fit the different wire rope diameters. This is a heavy duty stop sleeve made in the USA that provides a secure end termination for wire rope.