Learn More
Grainger Wire Rope Stop Sleeve 3/16 in SS
Catalog No. 502765638
Encompass_Preferred
The Loos Wire Rope Stop Sleeve Stop is a wire rope termination device designed to prevent fraying and unraveling of wire rope ends. It is made from drop-forged steel with a zinc finish and has a working load limit of 1 ton. This wire rope stop can accommodate wire rope diameters from 1/4" to 1/2". It secures the wire rope by compressing it when installed. Overall length is 2-3/4", width is 1-1/4", diameter is 1". This wire rope stop provides a finished look and protects against accidents. It is recommended for overhead lifting applications.
Catalog No. 50-276-5638 Supplier Grainger Supplier No. 16X638
The Loos Wire Rope Stop Sleeve Stop is a wire rope termination device designed to prevent fraying and unraveling of wire rope ends. It is made from drop-forged steel with a zinc finish and has a working load limit of 1 ton. This wire rope stop can accommodate wire rope diameters from 1/4" to 1/2". It secures the wire rope by compressing it when installed. Overall length is 2-3/4", width is 1-1/4", diameter is 1". This wire rope stop provides a finished look and protects against accidents. It is recommended for overhead lifting applications.