Learn More
Grainger Wire Rope Stud 1/2 -20 RH Thread 5/16 in
Catalog No. 502765114
Encompass_Preferred
The Grainger 9/16"-18 Swage Stud Fittings for Wire Rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged collar design that grips the wire rope for a secure connection. The fittings have a working load limit of 5,400 lbs and are zinc plated for corrosion resistance. They are available in a pack of 10 fittings with dimensions of 1-1/4" diameter x 2-1/2" length. The fittings meet military specification MIL-S-21433 and are recommended for general purpose applications with 9/16" diameter wire rope.
Catalog No. 50-276-5114 Supplier Grainger Supplier No. 16X705
The Grainger 9/16"-18 Swage Stud Fittings for Wire Rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged collar design that grips the wire rope for a secure connection. The fittings have a working load limit of 5,400 lbs and are zinc plated for corrosion resistance. They are available in a pack of 10 fittings with dimensions of 1-1/4" diameter x 2-1/2" length. The fittings meet military specification MIL-S-21433 and are recommended for general purpose applications with 9/16" diameter wire rope.