Learn More
Grainger Wire Rope Stud 1/4 -28 LH Thread 5/32 in
Catalog No. 502767771
Encompass_Preferred
The Grainger 16X695 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This UL rated, 2.5 lb stainless steel extinguisher uses Novec 1230 fluid which leaves no residue and is ideal for protecting valuable equipment. It features a steel cylinder with red powder coat finish, 12" hose, and pistol grip handle for easy operation. The extinguisher has a discharge range of 9-15 ft and is rechargeable. It meets NFPA 2010 requirements. Overall, this is an effective clean agent fire extinguisher well-suited for electronic or industrial applications where residue-free suppression is needed.
Catalog No. 50-276-7771 Supplier Grainger Supplier No. 16X695
The Grainger 16X695 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This UL rated, 2.5 lb stainless steel extinguisher uses Novec 1230 fluid which leaves no residue and is ideal for protecting valuable equipment. It features a steel cylinder with red powder coat finish, 12" hose, and pistol grip handle for easy operation. The extinguisher has a discharge range of 9-15 ft and is rechargeable. It meets NFPA 2010 requirements. Overall, this is an effective clean agent fire extinguisher well-suited for electronic or industrial applications where residue-free suppression is needed.