Learn More
Grainger Wire Rope Stud 1/4 -28 RH Thread 1/8 in
Catalog No. 502768200
Encompass_Preferred
The Grainger 16X692 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-scratching fibers for cleaning and scouring. It has a 6 inch by 9 inch size and comes in a pack of 10 pads. The pads have a medium duty abrasiveness that is suitable for cleaning pots, pans, dishes, counters, walls, appliances, tools, and more. The open web design allows dirt and grime to collect in the pad for more effective scrubbing. It can be used for hand scrubbing or attaching to a handle. This pack of 10 general purpose hand pads from 3M and Grainger provides an abrasive scrubbing solution for various cleaning tasks.
Catalog No. 50-276-8200 Supplier Grainger Supplier No. 16X692
The Grainger 16X692 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-scratching fibers for cleaning and scouring. It has a 6 inch by 9 inch size and comes in a pack of 10 pads. The pads have a medium duty abrasiveness that is suitable for cleaning pots, pans, dishes, counters, walls, appliances, tools, and more. The open web design allows dirt and grime to collect in the pad for more effective scrubbing. It can be used for hand scrubbing or attaching to a handle. This pack of 10 general purpose hand pads from 3M and Grainger provides an abrasive scrubbing solution for various cleaning tasks.