Learn More
Grainger Wire Rope Stud 3/8 -24 LH Thread 1/4 in
Catalog No. 502765656
Encompass_Preferred
The Grainger 3/8"-24 swage stud fittings for wire rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged end that grips the wire rope for a secure connection. The stud end has coarse 3/8"-24 threads to accept various attachments. Load ratings vary by stud length from 5,400 lbs to 9,000 lbs. Available in multiple stud lengths from 1-1/4" to 3" for versatility. Zinc plating resists corrosion. Meets military, federal, and other specs. Quality US-made fittings for heavy duty wire rope applications.
Catalog No. 50-276-5656 Supplier Grainger Supplier No. 16X702
The Grainger 3/8"-24 swage stud fittings for wire rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged end that grips the wire rope for a secure connection. The stud end has coarse 3/8"-24 threads to accept various attachments. Load ratings vary by stud length from 5,400 lbs to 9,000 lbs. Available in multiple stud lengths from 1-1/4" to 3" for versatility. Zinc plating resists corrosion. Meets military, federal, and other specs. Quality US-made fittings for heavy duty wire rope applications.