Learn More
Grainger Wire Rope Stud 3/8 -24 RH Thread 7/32 in
Catalog No. 502767773
Encompass_Preferred
The LOOS Wire Rope Stud is a 3/32 inch wire rope stud that is compatible with 3/32 inch wire rope. It is made from galvanized steel and has a working load limit of 75 pounds. The stud has a head diameter of 0.375 inches and an overall length of 0.5 inches. It can be used for creating an endless wire rope sling by inserting the wire rope and swaging. This wire rope stud is ideal for light duty applications.
Catalog No. 50-276-7773 Supplier Grainger Supplier No. 16X698
The LOOS Wire Rope Stud is a 3/32 inch wire rope stud that is compatible with 3/32 inch wire rope. It is made from galvanized steel and has a working load limit of 75 pounds. The stud has a head diameter of 0.375 inches and an overall length of 0.5 inches. It can be used for creating an endless wire rope sling by inserting the wire rope and swaging. This wire rope stud is ideal for light duty applications.