Learn More
Grainger Wire Rope Stud 3/8 -24 RH Thread 1/4 in
Catalog No. 502768237
Encompass_Preferred
The Grainger Approved Wire Rope Stud is a galvanized steel wire rope stud with a 1/2" diameter and 16" overall length. It has a working load limit of 1,650 lbs and is designed for general purpose use in wire rope applications. The stud features a forged eyelet on one end and is threaded on the other end to accept a nut. It can be used to create an endless wire rope sling or as an end termination. This wire rope stud is galvanized for corrosion resistance. With its high working load and corrosion resistant finish, this wire rope stud is a versatile option for creating wire rope assemblies.
Catalog No. 50-276-8237 Supplier Grainger Supplier No. 16X701
The Grainger Approved Wire Rope Stud is a galvanized steel wire rope stud with a 1/2" diameter and 16" overall length. It has a working load limit of 1,650 lbs and is designed for general purpose use in wire rope applications. The stud features a forged eyelet on one end and is threaded on the other end to accept a nut. It can be used to create an endless wire rope sling or as an end termination. This wire rope stud is galvanized for corrosion resistance. With its high working load and corrosion resistant finish, this wire rope stud is a versatile option for creating wire rope assemblies.