Learn More
Grainger Wire Rope Stud 5/16 -24 RH Thread 3/16in
Catalog No. 502768233
Encompass_Preferred
The Loos Wire Rope Stud is a 3/32 inch wire rope stud that is compatible with Loos tensioners. It is made from stainless steel for durability and corrosion resistance. The stud has a working load limit of 550 lbs and ultimate breaking strength of 1,650 lbs. It can be used for a variety of rigging and wire rope termination applications. The stud meets military specification MIL-S-781. It comes in a package of 100 pieces.
Catalog No. 50-276-8233 Supplier Grainger Supplier No. 16X696
The Loos Wire Rope Stud is a 3/32 inch wire rope stud that is compatible with Loos tensioners. It is made from stainless steel for durability and corrosion resistance. The stud has a working load limit of 550 lbs and ultimate breaking strength of 1,650 lbs. It can be used for a variety of rigging and wire rope termination applications. The stud meets military specification MIL-S-781. It comes in a package of 100 pieces.