Learn More
Grainger Wire Rope Stud #6-40 LH Thread 1/16 in
Catalog No. 502765113
Encompass_Preferred
The Grainger 1/16" Wire Rope Stud has a left hand thread, is made of carbon steel, and has a working load limit of 90 pounds. It can be used for commercial or industrial applications. The overall length is 1-3/4" and it weighs 0.01 lbs. This wire rope stud is zinc plated for corrosion resistance. It meets Federal Specification FF-S-325 Group II, Type I Class 3.
Catalog No. 50-276-5113 Supplier Grainger Supplier No. 16X689
The Grainger 1/16" Wire Rope Stud has a left hand thread, is made of carbon steel, and has a working load limit of 90 pounds. It can be used for commercial or industrial applications. The overall length is 1-3/4" and it weighs 0.01 lbs. This wire rope stud is zinc plated for corrosion resistance. It meets Federal Specification FF-S-325 Group II, Type I Class 3.