Learn More
Grainger Wire Rope Stud 9/16 -18 LH Thread 3/8 in
Catalog No. 502767087
Encompass_Preferred
The Grainger 9/16"-18 Swage Stud Fittings for Wire Rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged collar design that grips the wire rope for a strong and secure connection. The fittings have a working load limit of 5,400 lbs and are rated for 6x7 and 6x19 class wire rope constructions. They are zinc plated for corrosion resistance. The fittings accept 9/16"-18 threaded studs and are available in a range of sizes to accommodate different wire rope diameters from 3/16" to 1/2". Overall, these swage stud fittings provide a reliable threaded end termination for wire rope in lifting and rigging applications.
Catalog No. 50-276-7087 Supplier Grainger Supplier No. 16X708
The Grainger 9/16"-18 Swage Stud Fittings for Wire Rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged collar design that grips the wire rope for a strong and secure connection. The fittings have a working load limit of 5,400 lbs and are rated for 6x7 and 6x19 class wire rope constructions. They are zinc plated for corrosion resistance. The fittings accept 9/16"-18 threaded studs and are available in a range of sizes to accommodate different wire rope diameters from 3/16" to 1/2". Overall, these swage stud fittings provide a reliable threaded end termination for wire rope in lifting and rigging applications.