Learn More
Grainger Wire Rope Stud 9/16 -18 RH Thread 3/8 in
Catalog No. 502768041
Encompass_Preferred
The Grainger 9/16"-18 swage stud fittings for wire rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged collar design that grips the wire rope for a strong and secure connection. The fittings have a working load limit of 5,400 lbs and are rated for 6x19 and 6x37 class wire rope. They are zinc plated for corrosion resistance. The fittings accept 9/16"-18 threaded studs and are available in a range of sizes to accommodate different wire rope diameters from 3/8" to 1-1/4". Overall, these swage stud fittings provide a reliable threaded end termination for wire rope in lifting and rigging applications.
Catalog No. 50-276-8041 Supplier Grainger Supplier No. 16X707
The Grainger 9/16"-18 swage stud fittings for wire rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged collar design that grips the wire rope for a strong and secure connection. The fittings have a working load limit of 5,400 lbs and are rated for 6x19 and 6x37 class wire rope. They are zinc plated for corrosion resistance. The fittings accept 9/16"-18 threaded studs and are available in a range of sizes to accommodate different wire rope diameters from 3/8" to 1-1/4". Overall, these swage stud fittings provide a reliable threaded end termination for wire rope in lifting and rigging applications.