Learn More
Uline Wire Security Cage - 60 x 24 x 63""""
Supplier:  Uline H8397
Encompass
Welded steel wire cage, 60 x 24 x 63"""", 2 heavy duty shelves, lockable double doors, shelf leveling footplates for uneven floors, anchor to floor for extra stability with included hardware, capacity = 650lbs per shelf
Catalog No. NC2540424