Learn More
Uline Epoxy Wire Shelving Unit - 60 x 24 x 72", Zinc
Supplier:  Uline H3820
Encompass
Coated shelving provides superior chemical resistance.Recommended for wet environments, labs and kitchen coolers.Epoxy coating withstands harsh cleaning, salt and water.Shelves adjust in 1" increments.NSF certified.Optional 5" polyurethane swivel stem casters add 6" to height.
Catalog No. NC2865782