Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Wire Shlv Unit Cap 600 lb 24in

Supplier:  Grainger 1ANZ8

Encompass_Preferred

Wire Shlv Unit Cap 600 lb 24inx74inx36in

Catalog No. 50-279-2927


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.