Learn More
World Precision Instrument Wire Speculum, 4.5cm, 13.5mm Blades, Titanium
SDP
Catalog No. 502538945
Encompass
A wire eyelid speculum is a surgical instrument used by ophthalmologists to keep eyelids open during an exam or surgery. It consists of two curved blades that are gently inserted between the upper and lower eyelids to hold them open, allowing the ophthalmologist a clear view of the eye. Our wire speculum is made of titanium, which is non-magnetic and lightweight. It has a maximum spread of 20mm. This wire speculum can be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect and sterilize your surgical instruments promptly after each use to ensure they last for many years.
Catalog No. 50-253-8945 Supplier World Precision Instrument Supplier No. 555801L
A wire eyelid speculum is a surgical instrument used by ophthalmologists to keep eyelids open during an exam or surgery. It consists of two curved blades that are gently inserted between the upper and lower eyelids to hold them open, allowing the ophthalmologist a clear view of the eye. Our wire speculum is made of titanium, which is non-magnetic and lightweight. It has a maximum spread of 20mm. This wire speculum can be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect and sterilize your surgical instruments promptly after each use to ensure they last for many years.