Learn More
World Precision Instrument Wire Speculum, 4cm, 5x5mm Wire Blades
SDP
Catalog No. 502521361
Encompass
A wire eyelid speculum is a surgical instrument used by ophthalmologists to keep eyelids open during an exam or surgery. It consists of two curved blades that are gently inserted between the upper and lower eyelids to hold them open, allowing the ophthalmologist a clear view of the eye. Our 4cm wire specula are made of surgical grade stainless steel, and they come in two sizes, with a spread of 15mm. Choose the dimensions of your blades: 5 x 5mm or 5 x 13mm. This wire speculum can be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect and sterilize your surgical instruments promptly after each use to ensure they last for many years.
Catalog No. 50-252-1361 Supplier World Precision Instrument Supplier No. 501993
A wire eyelid speculum is a surgical instrument used by ophthalmologists to keep eyelids open during an exam or surgery. It consists of two curved blades that are gently inserted between the upper and lower eyelids to hold them open, allowing the ophthalmologist a clear view of the eye. Our 4cm wire specula are made of surgical grade stainless steel, and they come in two sizes, with a spread of 15mm. Choose the dimensions of your blades: 5 x 5mm or 5 x 13mm. This wire speculum can be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect and sterilize your surgical instruments promptly after each use to ensure they last for many years.