Learn More
Med Vet International Wire Test Tube Rack, J0352, 6in x 12in, Holds 72 Tubes, Each
Catalog No. 5000584649
Encompass_Preferred
Tough. Corrosion-resistant. Epoxy coated. Three-layered deck with square holes. Holds 72 tubes in a 6 x 12 configuration.Measurements: 2 5/8" tall x 5 1/8" wide x 9 1/2" long.
Catalog No. 50-005-84649 Supplier Med Vet International Supplier No. JOR352
Tough. Corrosion-resistant. Epoxy coated. Three-layered deck with square holes. Holds 72 tubes in a 6 x 12 configuration.Measurements: 2 5/8" tall x 5 1/8" wide x 9 1/2" long.