Learn More
Medicus Health WIRE TUBE RACKS, 8 PLACE, FOR 50 ML TEST TUBES
SDP
Catalog No. NC1915403
Encompass
∙ Wire Tube Racks with Epoxy Coating are Strong and Durable Solutions for Tough Lab Conditions∙50 mL rack has 30 mm square holes∙Chemical-resistant, epoxy-coated steel racks feature a slotted bottom and reinforced edges∙Ideal for conical centrifuge tubes but can be used to hold other types and sizes of tubes∙Tubes sit securely in the square holes and the middle rung provides added support∙Autoclavable for easy clean up
Catalog No. NC1915403 Supplier Medicus Health Supplier No. 2280M1
∙ Wire Tube Racks with Epoxy Coating are Strong and Durable Solutions for Tough Lab Conditions∙50 mL rack has 30 mm square holes∙Chemical-resistant, epoxy-coated steel racks feature a slotted bottom and reinforced edges∙Ideal for conical centrifuge tubes but can be used to hold other types and sizes of tubes∙Tubes sit securely in the square holes and the middle rung provides added support∙Autoclavable for easy clean up