Learn More
Med Vet International Wire Twister, 7" long, J0215L, Each
Catalog No. 5000585561
Encompass_Preferred
Will work with any size wire. For cerclage, hemicerglage and tension band wiring. Stainless steel.
Catalog No. 50-005-85561 Supplier Med Vet International Supplier No. JOR215L
Will work with any size wire. For cerclage, hemicerglage and tension band wiring. Stainless steel.