Learn More
Med Vet International Wire Twister/Shear Cutter 15cm, J0218, Each
Catalog No. 5000585488
Encompass_Preferred
Holds and twists all gauges of orthopedic wire. Cuts wire up to 18g twisted double. Ratchet locks jaws onto wire. Cutter has shear action for long life. Small hole in jaws threads wire for secure grip.
Catalog No. 50-005-85488 Supplier Med Vet International Supplier No. JOR218
Holds and twists all gauges of orthopedic wire. Cuts wire up to 18g twisted double. Ratchet locks jaws onto wire. Cutter has shear action for long life. Small hole in jaws threads wire for secure grip.