Learn More
Innovive Inc Wirebar, Rat, SS
Catalog No. NC1178731
Encompass
Rat wire bar protects the water bottle and holds food. Stainless steel reusable wire bar. For use with Innocage Standard Rat bottom.
Catalog No. NC1178731 Supplier Innovive Inc Supplier No. RWIREHB
Rat wire bar protects the water bottle and holds food. Stainless steel reusable wire bar. For use with Innocage Standard Rat bottom.