Learn More
MSC Milwaukee Wireless Dust Control Adapter & Remote Kit, Corded Vacuum, Quick Pairing, Single Outlet
Catalog No. 502838668
Encompass
The Wireless Dust Control Adapter & Remote Kit provides instant dust control and reduces downtime by allowing remote operation of any corded vacuum or dust extractor. The remote pairs quickly with the adapter, and the adapter's compact design ensures it uses only one spot in a standard outlet, leaving the other accessible.
  • Designed for use with any corded vacuum or dust extractor.
  • Pairs to the adapter in 3 seconds or less.
  • Multiple remotes can be paired to the same adapter.
  • Features a compact, lightweight design.
  • Equipped with LEDs that indicate pairing and function.
  • Adapter uses only one spot in a standard outlet.
  • Remote includes an adjustable strap and removable belt clip.
  • Remote has a 100-foot range.
Catalog No. 50-283-8668 Supplier MSC Supplier No. 83128280
The Wireless Dust Control Adapter & Remote Kit provides instant dust control and reduces downtime by allowing remote operation of any corded vacuum or dust extractor. The remote pairs quickly with the adapter, and the adapter's compact design ensures it uses only one spot in a standard outlet, leaving the other accessible.
  • Designed for use with any corded vacuum or dust extractor.
  • Pairs to the adapter in 3 seconds or less.
  • Multiple remotes can be paired to the same adapter.
  • Features a compact, lightweight design.
  • Equipped with LEDs that indicate pairing and function.
  • Adapter uses only one spot in a standard outlet.
  • Remote includes an adjustable strap and removable belt clip.
  • Remote has a 100-foot range.