Learn More
Research Products International Corp Wireless Humidity/Temperature Monitor Set
SDP
Catalog No. 501367784
Encompass
Monitor relative humidity and temperature inside an environmental chamber or clean room without opening the door. Place the weatherproof sensor outdoors, in green houses, warehouses, air ducts and other hard to reach places and continuously monitor ambient relative humidity and temperature from a remote location.
The wireless sensors broadcast to the monitor from up to 100 feet (30 M) away without obstructions, using a wireless RF signal. The monitor displays the local and remote temperatures in ºC or ºF, recalls min/max on both, and indicates when the batteries are low in both the sensors and monitor. It also displays user adjustable, audible and visible, Hi / Lo Alarms, on both temperature and relative humidity, for the base unit and the remote sensors.
Supplied with 2 x AAA batteries for the monitor and 2 x AA batteries for the sensor.
Catalog No. 50-136-7784 Supplier Research Products International Corp Supplier No. 800254
Monitor relative humidity and temperature inside an environmental chamber or clean room without opening the door. Place the weatherproof sensor outdoors, in green houses, warehouses, air ducts and other hard to reach places and continuously monitor ambient relative humidity and temperature from a remote location.
The wireless sensors broadcast to the monitor from up to 100 feet (30 M) away without obstructions, using a wireless RF signal. The monitor displays the local and remote temperatures in ºC or ºF, recalls min/max on both, and indicates when the batteries are low in both the sensors and monitor. It also displays user adjustable, audible and visible, Hi / Lo Alarms, on both temperature and relative humidity, for the base unit and the remote sensors.
Supplied with 2 x AAA batteries for the monitor and 2 x AA batteries for the sensor.