Learn More
Perkin Elmer US LLC Wireless Router Kit for the Spectrum Two
Supplier:  Perkin Elmer US LLC L1600246
Encompass_Preferred
This router works with the Wireless Dongle (L9004198) to upload/download, share, and backup data gathered using the portable Spectrum Two N spectrometer. It comes standard in the Spectrum Two Portability Pack (L1600238), but can be ordered separately as a replacement or spare using this part number.
Catalog No. 50-211-7219