Learn More
H-B Instrument™ Wireless Weather Station
SCI_ED
Catalog No. S95625 Shop All H-B Instrument Company Products
This item is not returnable. View return policy
Catalog No. S95625 Supplier H-B Instrument™ Supplier No. B615000500
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Includes
Batteries and remote sensor
Become an amateur meteorologist with these all-in-one wireless stations.
  • Perpetual calendar up to year 2050 with date and day of week in seven languages (English, German, French, Italian, Spanish, Dutch, and Danish)
  • 12/24-hour selectable time with snooze alarm
  • Moon phase
  • Dual-scale indoor/outdoor temperature range — Indoor: 0°/50°C (35°/122°F); Outdoor: -20°/+60°C (-4°/+140°F)
  • Indoor/outdoor temperature trend indicator
  • RF 433MHz wireless thermometer links up to three remote sensors with 100 ft. transmission (30m)
  • Model 693 adds max./min. temperature memory and outdoor temperature alert
Order Info
Individually packaged with multi-language instructions.
Certifications
CE marked
Compliance
RoHS compliant
Specifications
Model 693: Weather Station Wireless
Includes Batteries and remote sensor