Learn More
Grainger Wiring Harness 10 ft.
Catalog No. 502766179
Encompass_Preferred
The Edwards Signaling 10 ft Wiring Harness is a 10 foot long wiring harness for fire alarm systems. It is made of 18 AWG wire with a PVC jacket and has 10 conductors. The harness has plugs on both ends that allow it to easily connect devices like pull stations, smoke detectors, horns and strobes. It can handle up to 600 volts and meet UL and NEC standards. The harness provides a pre-wired solution for installing alarm devices and reduces installation time.
Catalog No. 50-276-6179 Supplier Grainger Supplier No. 16X431
The Edwards Signaling 10 ft Wiring Harness is a 10 foot long wiring harness for fire alarm systems. It is made of 18 AWG wire with a PVC jacket and has 10 conductors. The harness has plugs on both ends that allow it to easily connect devices like pull stations, smoke detectors, horns and strobes. It can handle up to 600 volts and meet UL and NEC standards. The harness provides a pre-wired solution for installing alarm devices and reduces installation time.