Learn More
Grainger Wiring Harness 5 ft.
Catalog No. 502766941
Encompass_Preferred
The Grainger 16X433 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1/2" diameter shaft and 1-1/4" long. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It operates on 115V, 60Hz single phase power and draws 8.6 full load amps. This general purpose motor can be used for pumps, fans, compressors, conveyors, and other industrial applications requiring 1/2 HP.
Catalog No. 50-276-6941 Supplier Grainger Supplier No. 16X433
The Grainger 16X433 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1/2" diameter shaft and 1-1/4" long. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It operates on 115V, 60Hz single phase power and draws 8.6 full load amps. This general purpose motor can be used for pumps, fans, compressors, conveyors, and other industrial applications requiring 1/2 HP.