Learn More
Cayman Chemical Wiskostatin, Purity: ≥98%, CAS: 253449-04-6, 50 mg
SDP
Catalog No. 502972254
Encompass_Preferred
A cell-permeable, selective inhibitor of N-WASP (IC50 = ~10 μM); directly associates with the GTPase-binding domain of N-WASP, preventing its activation by Cdc42 and PIP2; interferes with membrane transport, endocytosis, and neurological development
Catalog No. 50-297-2254 Supplier Cayman Chemical Supplier No. 1504750 MG
A cell-permeable, selective inhibitor of N-WASP (IC50 = ~10 μM); directly associates with the GTPase-binding domain of N-WASP, preventing its activation by Cdc42 and PIP2; interferes with membrane transport, endocytosis, and neurological development