Learn More
Abcam Wiskostatin, N-WASP inhibitor, 10MG
Catalog No. 502706920
Encompass_Preferred
MW 426.1 Da, Purity >99%. Selective neural Wiskott-Aldrich syndrome protein (N-WASP) inhibitor. Binds the GTPase binding domain stabilising the autoinhibited conformation. Prevents activation of Arp2/3 complex in the actin cytoskeleton.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6920 Supplier Abcam Supplier No. AB14108510MG
MW 426.1 Da, Purity >99%. Selective neural Wiskott-Aldrich syndrome protein (N-WASP) inhibitor. Binds the GTPase binding domain stabilising the autoinhibited conformation. Prevents activation of Arp2/3 complex in the actin cytoskeleton.
The product is subject to the following: Abcam Restricted Use Statement