Learn More
Bellco Glass, Inc. WISP S/T Clr Glass Vial,4.0mL 15x45mm
SDP
Catalog No. 501945363
Encompass_Preferred
Designed for use with Shimadzu aoc-14/1400 and waters 48 model auto-samplers. MADE IN THE USA. Type 1 class a Borosilicate glass Size: 15x45mm
Catalog No. 50-194-5363 Supplier Bellco Glass, Inc. Supplier No. 207000090
Designed for use with Shimadzu aoc-14/1400 and waters 48 model auto-samplers. MADE IN THE USA. Type 1 class a Borosilicate glass Size: 15x45mm