Learn More
IPHASE PHARMA SERVICES LLC Wistar Han Rat Liver Microsomes,Female,0.5mL,20mg/mL
Catalog No. NC3860801
Encompass_Preferred
A major tool for metabolism stability study. Our liver microsomes products available in multi-species (human, monkey, beagle, rat, and mouse). We also offer customized products prepared from unconventional animal species, disease models, or animals of certain ages.
Catalog No. NC3860801 Supplier IPHASE PHARMA SERVICES LLC Supplier No. 0121D3.02
A major tool for metabolism stability study. Our liver microsomes products available in multi-species (human, monkey, beagle, rat, and mouse). We also offer customized products prepared from unconventional animal species, disease models, or animals of certain ages.