Learn More
Creative Biogene WISTAR MESENCHYM STEM CELL 1VL
SDP
Catalog No. NC3803542
Encompass
Wistar mesenchymal stem cells were taken from the inguinal fat of Wistar rats. These cells can express specific proteins of ADSCs and have strong proliferation and multidirectional differentiation capabilities. It can be used as a cell model to study proliferation aging immunity differentiation and transplantation.
Catalog No. NC3803542 Supplier Creative Biogene Supplier No. CSCC1297
Add to Cart
Add to Cart
Wistar mesenchymal stem cells were taken from the inguinal fat of Wistar rats. These cells can express specific proteins of ADSCs and have strong proliferation and multidirectional differentiation capabilities. It can be used as a cell model to study proliferation aging immunity differentiation and transplantation.