Learn More
iQ Biosciences Wistar Rat Bone Marrow Cells, 20M cells/vial
Catalog No. NC1463576
Encompass_Preferred
High-quality, high-viability (typically > 70%) bone marrow cells from the Wistar rat, a widely-used outbred rat strain. Used for a wide variety of immunology-based applications, including cell population characterization and differentiation into specific cell types. All orders come with an iQ Certificate of Analysis.
Catalog No. NC1463576 Supplier iQ Biosciences Supplier No. IQBRBM202
High-quality, high-viability (typically > 70%) bone marrow cells from the Wistar rat, a widely-used outbred rat strain. Used for a wide variety of immunology-based applications, including cell population characterization and differentiation into specific cell types. All orders come with an iQ Certificate of Analysis.